siRNA Details
Gene Symbol: DBF4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025104.3 | NP_079380.2 | 10 | 953 | ||
NM_145663.2 | NP_663696.1 | 10 | 953 | |||
NR_036623.1 | 9 | 865 | ||||
XM_006722098.2 | XP_006722161.1 | 10 | 926 | |||
XM_006722102.3 | XP_006722165.1 | 8 | 828 | |||
XM_011525281.2 | XP_011523583.1 | 9 | 2067 | |||
XM_017025145.1 | XP_016880634.1 | 10 | 1067 | |||
XM_017025146.1 | XP_016880635.1 | 10 | 959 | |||
XM_017025147.1 | XP_016880636.1 | 9 | 1058 | |||
XM_017025149.1 | XP_016880638.1 | 10 | 1067 | |||
XM_017025151.1 | XP_016880640.1 | 10 | 1067 | |||
GenBank mRNA | AF448801.1 | - | 10 | 878 | ||
AF465820.1 | - | 4 | 615 | |||
BC016158.1 | - | 10 | 953 | 3536735 | ||
BC033660.2 | - | 9 | 787 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
