siRNA Details
Gene Symbol: RAB10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016131.4 | NP_057215.3 | 4, 5 | 1160 | ||
GenBank mRNA | AF086917.1 | - | 4, 5 | 505 | ||
AF106681.1 | - | 4, 5 | 501 | |||
AF297660.1 | - | 4, 5 | 411 | |||
AF498945.1 | - | 4, 5 | 411 | |||
AK023223.1 | - | 4, 5 | 910 | |||
AL517754.3 | - | 4, 5 | 921 | |||
BC000896.1 | - | 4, 5 | 486 | 3464547 | ||
CR457303.1 | - | 4, 5 | 411 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
