siRNA Details
Gene Symbol: RFX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000635.3 | NP_000626.2 | 18 | 4095 | ||
NM_134433.2 | NP_602309.1 | 17 | 4020 | |||
XM_011528171.2 | XP_011526473.1 | 19 | 4061 | |||
XM_011528172.1 | XP_011526474.1 | 18 | 3922 | |||
XM_017027107.1 | XP_016882596.1 | 17 | 3945 | |||
XM_017027108.1 | XP_016882597.1 | 16 | 3870 | |||
GenBank mRNA | BC071571.1 | - | 18 | 3976 | 5276369 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
