siRNA Details
Gene Symbol: SPTSSB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040100.1 | NP_001035189.1 | 3 | 913 | ||
NM_001320679.1 | NP_001307608.1 | 4 | 1919 | |||
XM_017005793.1 | XP_016861282.1 | 3 | 1953 | |||
XM_017005794.1 | XP_016861283.1 | 4 | 2092 | |||
GenBank mRNA | AF458592.1 | - | 3 | 796 | ||
AK123348.1 | - | 3 | 1710 | |||
AK312144.1 | - | 1 | 168 | |||
BC037817.2 | - | 3 | 790 | |||
BC065208.1 | - | 3 | 337 | 5207108 | ||
BC071875.1 | - | 3 | 326 | |||
BC107756.1 | - | 3 | 351 | 6737276 | ||
CA487194.1 | - | 4 | 470 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
