siRNA Details
Gene Symbol: SPTSSB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040100.1 | NP_001035189.1 | 3 | 940 | ||
NM_001320679.1 | NP_001307608.1 | 4 | 1946 | |||
XM_017005793.1 | XP_016861282.1 | 3 | 1980 | |||
XM_017005794.1 | XP_016861283.1 | 4 | 2119 | |||
GenBank mRNA | AF458592.1 | - | 3 | 823 | ||
AK123348.1 | - | 3 | 1737 | |||
AK312144.1 | - | 1 | 195 | |||
BC037817.2 | - | 3 | 817 | |||
BC065208.1 | - | 3 | 364 | 5207108 | ||
BC071875.1 | - | 3 | 353 | |||
BC107756.1 | - | 3 | 378 | 6737276 | ||
CA487194.1 | - | 4 | 497 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
