siRNA Details
Gene Symbol: HIST1H2AG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC016677.1 | - | 1 | 730 | 4342191 | |
L19778.1 | - | 1 | 741 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
