siRNA Details
Gene Symbol: ELAVL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001420.3 | NP_001411.2 | 3 | 634 | ||
NM_032281.2 | NP_115657.2 | 3 | 634 | |||
XM_011527778.2 | XP_011526080.1 | 3 | 637 | |||
GenBank mRNA | AY034002.1 | - | 2 | 223 | ||
BC014144.2 | - | 3 | 627 | |||
D26158.1 | - | 3 | 500 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
