siRNA Details
Gene Symbol: ELAVL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001420.3 | NP_001411.2 | 7 | 3038 | ||
NM_032281.2 | NP_115657.2 | 7 | 3017 | |||
GenBank mRNA | BC011875.2 | - | 1 | 785 | 3161837 | |
BC080533.1 | - | 1 | 783 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
