siRNA Details
Gene Symbol: ELAVL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001420.3 | NP_001411.2 | 7 | 2200 | ||
NM_032281.2 | NP_115657.2 | 7 | 2179 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
