siRNA Details
Gene Symbol: PHF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005392.3 | NP_005383.3 | 21, 22 | 3360 | ||
XM_005252051.2 | XP_005252108.1 | 21, 22 | 3357 | |||
XM_006717143.2 | XP_006717206.1 | 21, 22 | 3252 | |||
GenBank mRNA | AB014562.2 | - | 20, 21 | 3268 | ||
AK294861.1 | - | 9, 10 | 1516 | |||
AL834263.1 | - | 21, 22 | 3226 | |||
BC050112.1 | - | 2, 3 | 592 | |||
BC064613.1 | - | 8, 9 | 948 | |||
BC152437.1 | - | 20, 21 | 3268 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
