siRNA Details
Gene Symbol: PHF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005392.3 | NP_005383.3 | 22 | 5293 | ||
XM_005252051.2 | XP_005252108.1 | 22 | 5290 | |||
XM_006717143.2 | XP_006717206.1 | 22 | 5185 | |||
GenBank mRNA | AB014562.2 | - | 21 | 5201 | ||
AL834263.1 | - | 22 | 5160 | |||
BC028051.2 | - | 10 | 1562 | |||
BC064613.1 | - | 9 | 2881 | |||
BC152437.1 | - | 21 | 5201 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
