siRNA Details
Gene Symbol: NF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000268.3 | NP_000259.1 | 14 | 2007 | ||
NM_016418.5 | NP_057502.2 | 14 | 2007 | |||
NM_181825.2 | NP_861546.1 | 14 | 2007 | |||
NM_181828.2 | NP_861966.1 | 13 | 1881 | |||
NM_181829.2 | NP_861967.1 | 13 | 1884 | |||
NM_181830.2 | NP_861968.1 | 12 | 1758 | |||
NM_181831.2 | NP_861969.1 | 12 | 1758 | |||
NM_181832.2 | NP_861970.1 | 14 | 2007 | |||
XM_017028809.1 | XP_016884298.1 | 14 | 1570 | |||
XM_017028810.1 | XP_016884299.1 | 14 | 1566 | |||
GenBank mRNA | AF122828.1 | - | 7 | 796 | ||
AF123570.1 | - | 9 | 830 | |||
AF369657.1 | - | 14 | 1989 | |||
AF369658.1 | - | 14 | 1989 | |||
AF369661.1 | - | 14 | 1564 | |||
AF369662.1 | - | 14 | 1564 | |||
AF369663.1 | - | 13 | 1438 | |||
AF369664.1 | - | 13 | 1441 | |||
AF369665.1 | - | 12 | 1315 | |||
AF369666.1 | - | 15 | 1680 | |||
AF369667.1 | - | 14 | 1589 | |||
AF369668.1 | - | 14 | 1564 | |||
AF369700.1 | - | 6 | 631 | |||
AF369701.1 | - | 14 | 1930 | |||
AK297116.1 | - | 14 | 1871 | |||
BC003112.1 | - | 12 | 1587 | 3537045 | ||
BC020257.1 | - | 14 | 1902 | 4871980 | ||
CR456530.1 | - | 14 | 1626 | |||
L11353.1 | - | 14 | 1783 | |||
Z22664.1 | - | 14 | 1738 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
