siRNA Details
Gene Symbol: DDX42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007372.3 | NP_031398.2 | 7, 8 | 902 | ||
NM_203499.2 | NP_987095.1 | 6, 7 | 831 | |||
XM_006721657.1 | XP_006721720.1 | 7, 8 | 1039 | |||
XM_017024111.1 | XP_016879600.1 | 8, 9 | 1138 | |||
XM_017024112.1 | XP_016879601.1 | 2, 3 | 218 | |||
GenBank mRNA | AB036090.1 | - | 5, 6 | 627 | ||
AF083255.1 | - | 4, 5 | 287 | |||
AK292908.1 | - | 4, 5 | 414 | |||
BC015505.1 | - | 5, 6 | 612 | 3905607 | ||
BC078667.1 | - | 5, 6 | 570 | |||
BC093081.1 | - | 7, 8 | 866 | 30407786 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
