siRNA Details
Gene Symbol: DDX42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007372.3 | NP_031398.2 | 16 | 2019 | ||
NM_203499.2 | NP_987095.1 | 15 | 1948 | |||
XM_006721657.1 | XP_006721720.1 | 16 | 2156 | |||
XM_011524259.2 | XP_011522561.1 | 10 | 1957 | |||
XM_017024111.1 | XP_016879600.1 | 17 | 2255 | |||
XM_017024112.1 | XP_016879601.1 | 11 | 1335 | |||
GenBank mRNA | AB036090.1 | - | 14 | 1744 | ||
AF083255.1 | - | 13 | 1404 | |||
AK001964.1 | - | 2 | 229 | |||
AK122737.1 | - | 2 | 998 | |||
AK126480.1 | - | 8 | 2046 | |||
AK292908.1 | - | 13 | 1531 | |||
AL050096.1 | - | 1 | 25 | |||
BC015505.1 | - | 14 | 1729 | 3905607 | ||
BC078667.1 | - | 14 | 1687 | |||
BC093081.1 | - | 16 | 1983 | 30407786 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
