siRNA Details
Gene Symbol: FGF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000800.4 | NP_000791.1 | 4 | 1072 | ||
NM_001144892.2 | NP_001138364.1 | 4 | 592 | |||
NM_001144934.1 | NP_001138406.1 | 5 | 785 | |||
NM_001144935.1 | NP_001138407.1 | 4 | 691 | |||
NM_001257205.1 | NP_001244134.1 | 4 | 982 | |||
NM_001257206.1 | NP_001244135.1 | 4 | 979 | |||
NM_001257207.1 | NP_001244136.1 | 5 | 725 | |||
NM_001257208.1 | NP_001244137.1 | 4 | 723 | |||
NM_001257209.1 | NP_001244138.1 | 4 | 738 | |||
NM_001257210.1 | NP_001244139.1 | 4 | 722 | |||
NM_001257211.1 | NP_001244140.1 | 4 | 720 | |||
NM_001257212.1 | NP_001244141.1 | 4 | 589 | |||
NM_033136.3 | NP_149127.1 | 3 | 968 | |||
NM_033137.2 | NP_149128.1 | 2 | 426 | |||
XM_005268390.4 | XP_005268447.1 | 4 | 662 | |||
XM_011537607.2 | XP_011535909.1 | 5 | 782 | |||
GenBank mRNA | BC032697.1 | - | 4 | 589 | 5403677 | |
DA228737.1 | - | 3 | 539 | |||
DQ655917.2 | - | 4 | 964 | |||
M13361.1 | - | 3 | 536 | |||
X51943.1 | - | 3 | 533 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
