siRNA Details
Gene Symbol: C1orf116 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083924.1 | NP_001077393.1 | 3 | 3434 | ||
NM_023938.5 | NP_076427.2 | 4 | 3620 | |||
XM_005273259.1 | XP_005273316.1 | 2 | 3188 | |||
XM_006711530.1 | XP_006711593.1 | 4 | 3497 | |||
XM_011509973.2 | XP_011508275.1 | 3 | 3290 | |||
GenBank mRNA | AK096741.1 | - | 1 | 1236 | ||
BC001943.1 | - | 1 | 118 | |||
BC002325.2 | - | 1 | 118 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
