siRNA Details
Gene Symbol: PRKRA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001139517.1 | NP_001132989.1 | 7 | 1896 | ||
NM_001139518.1 | NP_001132990.1 | 8 | 1617 | |||
NM_001316362.1 | NP_001303291.1 | 9 | 1935 | |||
NM_003690.4 | NP_003681.1 | 8 | 1816 | |||
XM_011512063.1 | XP_011510365.1 | 8 | 1747 | |||
XM_011512066.1 | XP_011510368.1 | 9 | 1972 | |||
XM_017005159.1 | XP_016860648.1 | 9 | 1857 | |||
GenBank mRNA | AF072860.1 | - | 8 | 1714 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
