siRNA Details
Gene Symbol: CAGE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170692.1 | NP_001164163.1 | 9 | 2698 | ||
NM_001170693.1 | NP_001164164.1 | 9 | 2698 | |||
NM_205864.2 | NP_995586.1 | 7 | 2399 | |||
GenBank mRNA | AF414185.1 | - | 7 | 2398 | ||
AK097760.1 | - | 9 | 2397 | |||
BC026188.1 | - | 9 | 2371 | |||
BC026194.2 | - | 9 | 2438 | 4826784 | ||
BC033545.1 | - | 9 | 2438 | 4826548 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
