siRNA Details
Gene Symbol: MRPL33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004891.3 | NP_004882.1 | 4 | 267 | ||
NM_145330.2 | NP_663303.1 | 3 | 160 | |||
GenBank mRNA | AF047440.1 | - | 4 | 241 | ||
BC009475.1 | - | 4 | 219 | 4049053 | ||
BC020834.1 | - | 4 | 254 | 4775045 | ||
BC087846.1 | - | 3 | 124 | |||
BC090047.1 | - | 3 | 160 | 30393466 | ||
BI601959.1 | - | 4 | 270 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
