siRNA Details
Gene Symbol: HIST1H2AM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003514.2 | NP_003505.1 | 1 | 108 | ||
GenBank mRNA | AK312224.1 | - | 1 | 108 | ||
BC032686.1 | - | 1 | 123 | |||
BC032756.1 | - | 1 | 123 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
