siRNA Details
Gene Symbol: GPM6A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001261447.1 | NP_001248376.1 | 5 | 2221 | ||
NM_001261448.1 | NP_001248377.1 | 7 | 2529 | |||
NM_005277.4 | NP_005268.1 | 8 | 2630 | |||
NM_201591.2 | NP_963885.1 | 7 | 3112 | |||
NM_201592.2 | NP_963886.1 | 7 | 2571 | |||
NR_048571.1 | 6 | 2280 | ||||
XM_006714183.1 | XP_006714246.1 | 6 | 2200 | |||
XM_011531877.1 | XP_011530179.1 | 7 | 2469 | |||
XM_017008036.1 | XP_016863525.1 | 7 | 2523 | |||
GenBank mRNA | AK226176.1 | - | 7 | 2263 | ||
BC022508.1 | - | 7 | 2262 | 4798738 | ||
BC022528.1 | - | 8 | 2272 | 4799752 | ||
BC037853.1 | - | 3 | 2605 | |||
BC044612.1 | - | 7 | 2742 | 5259851 | ||
BC053954.1 | - | 7 | 2742 | |||
D49958.1 | - | 7 | 2317 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
