siRNA Details
Gene Symbol: GPM6A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001261447.1 | NP_001248376.1 | 5 | 1540 | ||
NM_001261448.1 | NP_001248377.1 | 7 | 1848 | |||
NM_005277.4 | NP_005268.1 | 8 | 1949 | |||
NM_201591.2 | NP_963885.1 | 7 | 2431 | |||
NM_201592.2 | NP_963886.1 | 7 | 1890 | |||
NR_048571.1 | 6 | 1599 | ||||
XM_006714183.1 | XP_006714246.1 | 6 | 1519 | |||
XM_011531877.1 | XP_011530179.1 | 7 | 1788 | |||
XM_017008036.1 | XP_016863525.1 | 7 | 1842 | |||
GenBank mRNA | AK226176.1 | - | 7 | 1582 | ||
BC022508.1 | - | 7 | 1581 | 4798738 | ||
BC022528.1 | - | 8 | 1591 | 4799752 | ||
BC037853.1 | - | 3 | 1924 | |||
BC044612.1 | - | 7 | 2061 | 5259851 | ||
BC053954.1 | - | 7 | 2061 | |||
D49958.1 | - | 7 | 1636 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
