siRNA Details
Gene Symbol: GPM6A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001261447.1 | NP_001248376.1 | 5 | 2708 | ||
NM_001261448.1 | NP_001248377.1 | 7 | 3016 | |||
NM_005277.4 | NP_005268.1 | 8 | 3117 | |||
NM_201591.2 | NP_963885.1 | 7 | 3599 | |||
NM_201592.2 | NP_963886.1 | 7 | 3058 | |||
NR_048571.1 | 6 | 2767 | ||||
XM_006714183.1 | XP_006714246.1 | 6 | 2687 | |||
XM_011531877.1 | XP_011530179.1 | 7 | 2956 | |||
XM_017008036.1 | XP_016863525.1 | 7 | 3010 | |||
GenBank mRNA | BC022508.1 | - | 7 | 2749 | 4798738 | |
BC022528.1 | - | 8 | 2759 | 4799752 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001253754, NM_001253756, NM_153581, BC023461, BC029711, and others.
Gene Ontology Categories:
