siRNA Details
Gene Symbol: SLC39A10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127257.1 | NP_001120729.1 | 4 | 1517 | ||
NM_020342.2 | NP_065075.1 | 4 | 1468 | |||
XM_005246689.4 | XP_005246746.2 | 4 | 1475 | |||
XM_011511504.2 | XP_011509806.1 | 4 | 1382 | |||
XM_011511505.2 | XP_011509807.1 | 5 | 1546 | |||
XM_011511506.2 | XP_011509808.1 | 4 | 4205 | |||
XM_011511507.2 | XP_011509809.1 | 4 | 1475 | |||
XM_017004522.1 | XP_016860011.1 | 4 | 2507 | |||
GenBank mRNA | AB033091.1 | - | 4 | 1381 | ||
AK291241.1 | - | 4 | 1380 | |||
AK294771.1 | - | 5 | 564 | |||
AL050294.1 | - | 4 | 1367 | |||
BC101516.1 | - | 4 | 1377 | 8069022 | ||
BC112223.1 | - | 4 | 1389 | 8327691 | ||
CR749813.1 | - | 4 | 1368 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
