siRNA Details
Gene Symbol: KLHL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007075.2 | NP_001007076.1 | 6 | 1290 | ||
NM_001171654.1 | NP_001165125.1 | 5 | 813 | |||
NM_015990.4 | NP_057074.3 | 5 | 1352 | |||
NM_199039.3 | NP_950240.2 | 4 | 1169 | |||
XM_005262655.4 | XP_005262712.1 | 6 | 1324 | |||
XM_011513699.2 | XP_011512001.1 | 6 | 1326 | |||
XM_011513700.2 | XP_011512002.1 | 6 | 1326 | |||
XM_011513701.2 | XP_011512003.1 | 6 | 1326 | |||
XM_017008273.1 | XP_016863762.1 | 6 | 1326 | |||
XM_017008274.1 | XP_016863763.1 | 6 | 1326 | |||
XM_017008275.1 | XP_016863764.1 | 6 | 1321 | |||
XM_017008276.1 | XP_016863765.1 | 6 | 1326 | |||
GenBank mRNA | AB208820.1 | - | 5 | 585 | ||
AF272976.1 | - | 5 | 1144 | |||
AK001698.1 | - | 3 | 342 | |||
AK002174.1 | - | 4 | 438 | |||
AK289785.1 | - | 5 | 1352 | |||
AY172948.1 | - | 4 | 961 | |||
BC053860.1 | - | 6 | 1092 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
