siRNA Details
Gene Symbol: OR5H6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005479.1 | NP_001005479.1 | 1 | 111 | ||
XM_011513118.2 | XP_011511420.1 | 2 | 157 | |||
GenBank mRNA | BC137523.1 | - | 1 | 152 | 9021523 | |
BC137524.1 | - | 1 | 152 | 9021524 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
