siRNA Details
Gene Symbol: MB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005368.2 | NP_005359.1 | 1 | 109 | ||
NM_203377.1 | NP_976311.1 | 2 | 201 | |||
NM_203378.1 | NP_976312.1 | 2 | 184 | |||
XM_005261605.2 | XP_005261662.1 | 2 | 145 | |||
GenBank mRNA | AK300657.1 | - | 1 | 101 | ||
AK314057.1 | - | 2 | 170 | |||
BC014547.1 | - | 1 | 105 | 4244838 | ||
BQ925917.1 | - | 233 | ||||
BQ956082.1 | - | 88 | ||||
CB140824.1 | - | 2 | 214 | |||
CR456516.1 | - | 1 | 77 | |||
CR541949.1 | - | 1 | 29 | |||
HG531366.1 | - | 1 | 67 | |||
HG531368.1 | - | 1 | 45 | |||
HG531369.1 | - | 1 | 44 | |||
HG531370.1 | - | 3 | 181 | |||
HG531372.1 | - | 2 | 193 | |||
HG531379.1 | - | 2 | 193 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
