siRNA Details
Gene Symbol: MB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005368.2 | NP_005359.1 | 3 | 798 | ||
NM_203377.1 | NP_976311.1 | 4 | 890 | |||
NM_203378.1 | NP_976312.1 | 4 | 873 | |||
XM_005261605.2 | XP_005261662.1 | 4 | 834 | |||
GenBank mRNA | AK300657.1 | - | 4 | 756 | ||
BC014547.1 | - | 3 | 794 | 4244838 | ||
BC018001.1 | - | 3 | 763 | 4271145 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
