siRNA Details
Gene Symbol: NUDT16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171905.1 | NP_001165376.1 | 4 | 1880 | ||
NM_152395.2 | NP_689608.2 | 3 | 2054 | |||
NR_033268.1 | 2 | 2141 | ||||
GenBank mRNA | AK055827.1 | - | 3 | 1956 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
