siRNA Details
Gene Symbol: ZNF182 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007088.1 | NP_001007089.1 | 6 | 1990 | ||
NM_001178099.1 | NP_001171570.1 | 7 | 2106 | |||
NM_006962.1 | NP_008893.1 | 7 | 2084 | |||
GenBank mRNA | AK097687.1 | - | 1 | 643 | ||
AK122874.1 | - | 7 | 2089 | |||
AK130362.1 | - | 1 | 1210 | |||
AK299325.1 | - | 6 | 1986 | |||
BC106875.2 | - | 6 | 1954 | 40011764 | ||
BC114216.1 | - | 6 | 1967 | 6069900 | ||
BX537686.1 | - | 1 | 975 | |||
BX648401.1 | - | 1 | 1306 | |||
X52345.1 | - | 1 | 115 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
