siRNA Details
Gene Symbol: LOC51145 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_122032.1 | 1 | 341 | |||
GenBank mRNA | AA397744.1 | - | 1 | 341 | ||
AF159054.1 | - | 1 | 964 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
