siRNA Details
Gene Symbol: ZNF182 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007088.1 | NP_001007089.1 | 6 | 2013 | ||
NM_001178099.1 | NP_001171570.1 | 7 | 2129 | |||
NM_006962.1 | NP_008893.1 | 7 | 2107 | |||
GenBank mRNA | AK097687.1 | - | 1 | 666 | ||
AK122874.1 | - | 7 | 2112 | |||
AK130362.1 | - | 1 | 1233 | |||
AK299325.1 | - | 6 | 2009 | |||
BC106875.2 | - | 6 | 1977 | 40011764 | ||
BC114216.1 | - | 6 | 1990 | 6069900 | ||
BX537686.1 | - | 1 | 998 | |||
BX648401.1 | - | 1 | 1329 | |||
X52345.1 | - | 1 | 138 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
