siRNA Details
Gene Symbol: ANKRD36B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025190.3 | NP_079466.3 | 3, 4 | 734 | ||
XM_005263995.4 | XP_005264052.1 | 3, 4 | 1014 | |||
XM_006712661.1 | XP_006712724.1 | 3, 4 | 1014 | |||
XM_011511555.1 | XP_011509857.1 | 3, 4 | 1014 | |||
XM_011511556.1 | XP_011509858.1 | 3, 4 | 1014 | |||
XM_011511557.2 | XP_011509859.1 | 3, 4 | 1014 | |||
XM_011511558.1 | XP_011509860.1 | 3, 4 | 1014 | |||
XM_017004578.1 | XP_016860067.1 | 3, 4 | 1014 | |||
XM_017004579.1 | XP_016860068.1 | 3, 4 | 1014 | |||
XM_017004580.1 | XP_016860069.1 | 3, 4 | 1014 | |||
XM_017004581.1 | XP_016860070.1 | 3, 4 | 1014 | |||
XM_017004582.1 | XP_016860071.1 | 3, 4 | 1014 | |||
XM_017004583.1 | XP_016860072.1 | 3, 4 | 1014 | |||
XM_017004584.1 | XP_016860073.1 | 3, 4 | 1014 | |||
XM_017004587.1 | XP_016860076.1 | 3, 4 | 1014 | |||
XM_017004588.1 | XP_016860077.1 | 3, 4 | 1014 | |||
XM_017004592.1 | XP_016860081.1 | 3, 4 | 1014 | |||
XM_017004593.1 | XP_016860082.1 | 3, 4 | 1014 | |||
GenBank mRNA | AK074570.1 | - | 3, 4 | 635 | ||
BC071771.1 | - | 3, 4 | 560 | |||
BM511319.1 | - | 3, 4 | 326 | |||
DR001190.1 | - | 3, 4 | 623 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
