siRNA Details
Gene Symbol: RPL34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000995.4 | NP_000986.2 | 6 | 522 | ||
NM_033625.3 | NP_296374.1 | 6 | 602 | |||
GenBank mRNA | BC001773.1 | - | 6 | 536 | 3355603 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
