siRNA Details
Gene Symbol: INE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024616.1 | 1 | 720 | |||
GenBank mRNA | BC069671.1 | - | 1 | 643 | ||
BC069696.1 | - | 1 | 642 | |||
BC069772.1 | - | 1 | 642 | |||
Y10696.1 | - | 1 | 646 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
