siRNA Details
Gene Symbol: CCNL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039577.3 | NP_001034666.1 | 1 | 289 | ||
NM_001320153.1 | NP_001307082.1 | 1 | 289 | |||
NM_001320155.1 | NP_001307084.1 | 1 | 289 | |||
NM_030937.4 | NP_112199.2 | 1 | 289 | |||
NR_135154.1 | 1 | 289 | ||||
XM_011542216.2 | XP_011540518.1 | 1 | 265 | |||
XM_017002420.1 | XP_016857909.1 | 1 | 287 | |||
XM_017002421.1 | XP_016857910.1 | 1 | 273 | |||
XM_017002422.1 | XP_016857911.1 | 1 | 286 | |||
XM_017002423.1 | XP_016857912.1 | 1 | 265 | |||
GenBank mRNA | AK056120.1 | - | 1 | 262 | ||
AK074112.1 | - | 1 | 260 | |||
AK225758.1 | - | 1 | 284 | |||
AL834441.1 | - | 1 | 220 | |||
AY037150.1 | - | 1 | 263 | |||
AY116620.1 | - | 1 | 278 | |||
BC016333.1 | - | 1 | 269 | |||
BM153748.1 | - | 1 | 289 | |||
DA360436.1 | - | 1 | 288 | |||
HY172534.1 | - | 1 | 289 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
