siRNA Details
Gene Symbol: INVS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318381.1 | NP_001305310.1 | 2 | 281 | ||
NM_001318382.1 | NP_001305311.1 | 2 | 281 | |||
NM_014425.4 | NP_055240.2 | 2 | 281 | |||
NR_134606.1 | 2 | 281 | ||||
GenBank mRNA | AF039217.2 | - | 2 | 232 | ||
AF084367.1 | - | 1 | 51 | |||
AK312795.1 | - | 2 | 223 | |||
BC006370.2 | - | 2 | 178 | |||
BC041665.1 | - | 2 | 237 | |||
BC056897.1 | - | 2 | 200 | |||
BC111761.1 | - | 1 | 64 | 40012263 | ||
BC112351.1 | - | 1 | 64 | 40012265 | ||
BP280580.1 | - | 2 | 246 | |||
DB217652.1 | - | 2 | 281 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
