siRNA Details
Gene Symbol: PRSS37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008270.2 | NP_001008271.2 | 3, 4 | 800 | ||
GenBank mRNA | AK172866.1 | - | 3, 4 | 800 | ||
BC137353.1 | - | 3, 4 | 590 | 9021351 | ||
BC137354.1 | - | 3, 4 | 590 | 9021352 | ||
BC144604.1 | - | 3, 4 | 558 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
