siRNA Details
Gene Symbol: PRSS37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008270.2 | NP_001008271.2 | 2, 3 | 540 | ||
NM_001171951.1 | NP_001165422.1 | 2, 3 | 540 | |||
XM_005249946.4 | XP_005250003.1 | 2, 3 | 8844 | |||
XM_005249947.3 | XP_005250004.1 | 3, 4 | 1662 | |||
XM_011515792.2 | XP_011514094.1 | 3, 4 | 666 | |||
GenBank mRNA | AK172866.1 | - | 2, 3 | 540 | ||
BC137353.1 | - | 2, 3 | 330 | 9021351 | ||
BC137354.1 | - | 2, 3 | 330 | 9021352 | ||
BC144604.1 | - | 2, 3 | 298 | |||
BC144605.1 | - | 2, 3 | 330 | 9053136 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
