siRNA Details
Gene Symbol: AFAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134647.1 | NP_001128119.1 | 2 | 379 | ||
NM_198595.2 | NP_940997.1 | 2 | 379 | |||
XM_006713909.3 | XP_006713972.1 | 2 | 262 | |||
XM_011513544.2 | XP_011511846.1 | 2 | 432 | |||
XM_017008535.1 | XP_016864024.1 | 1 | 108 | |||
XM_017008536.1 | XP_016864025.1 | 1 | 108 | |||
GenBank mRNA | AF188700.1 | - | 1 | 106 | ||
AK290807.1 | - | 2 | 206 | |||
AK297631.1 | - | 2 | 172 | |||
AL706541.1 | - | 1 | 324 | |||
BC032777.1 | - | 1 | 130 | 4914388 | ||
BI561136.1 | - | 2 | 379 | |||
DA605903.1 | - | 2 | 312 | |||
DB450895.1 | - | 2 | 379 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
