siRNA Details
Gene Symbol: AFAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134647.1 | NP_001128119.1 | 9 | 1234 | ||
NM_198595.2 | NP_940997.1 | 9 | 1234 | |||
XM_006713909.3 | XP_006713972.1 | 9 | 1117 | |||
XM_011513544.2 | XP_011511846.1 | 9 | 1287 | |||
XM_017008535.1 | XP_016864024.1 | 8 | 963 | |||
XM_017008536.1 | XP_016864025.1 | 8 | 963 | |||
GenBank mRNA | AB209676.1 | - | 6 | 687 | ||
AF188700.1 | - | 8 | 961 | |||
AK290807.1 | - | 9 | 1061 | |||
AK297631.1 | - | 9 | 1027 | |||
BC032777.1 | - | 8 | 985 | 4914388 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
