siRNA Details
Gene Symbol: PRKAR1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164758.1 | NP_001158230.1 | 8, 9 | 872 | ||
NM_001164759.1 | NP_001158231.1 | 8, 9 | 807 | |||
NM_001164760.1 | NP_001158232.1 | 8, 9 | 917 | |||
NM_001164761.1 | NP_001158233.1 | 8, 9 | 938 | |||
NM_001164762.1 | NP_001158234.1 | 8, 9 | 878 | |||
NM_002735.2 | NP_002726.1 | 8, 9 | 896 | |||
XM_011515446.2 | XP_011513748.1 | 7, 8 | 854 | |||
XM_017012433.1 | XP_016867922.1 | 3, 4 | 379 | |||
XM_017012434.1 | XP_016867923.1 | 5, 6 | 388 | |||
GenBank mRNA | AK098822.1 | - | 7, 8 | 838 | ||
AK289464.1 | - | 8, 9 | 819 | |||
AK297337.1 | - | 8, 9 | 848 | |||
AK315951.1 | - | 8, 9 | 908 | |||
AK315990.1 | - | 8, 9 | 868 | |||
AL833563.1 | - | 8, 9 | 838 | |||
BC026734.1 | - | 8, 9 | 846 | 5141110 | ||
BC036828.1 | - | 8, 9 | 869 | 5247772 | ||
BC070478.1 | - | 3, 4 | 360 | |||
M65066.1 | - | 7, 8 | 761 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
