siRNA Details
Gene Symbol: PRKAR1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164758.1 | NP_001158230.1 | 9 | 948 | ||
NM_001164759.1 | NP_001158231.1 | 9 | 883 | |||
NM_001164760.1 | NP_001158232.1 | 9 | 993 | |||
NM_001164761.1 | NP_001158233.1 | 9 | 1014 | |||
NM_001164762.1 | NP_001158234.1 | 9 | 954 | |||
NM_002735.2 | NP_002726.1 | 9 | 972 | |||
XM_011515446.2 | XP_011513748.1 | 8 | 930 | |||
XM_017012433.1 | XP_016867922.1 | 4 | 455 | |||
XM_017012434.1 | XP_016867923.1 | 6 | 464 | |||
GenBank mRNA | AK289464.1 | - | 9 | 895 | ||
AK297337.1 | - | 9 | 924 | |||
AK315951.1 | - | 9 | 984 | |||
AK315990.1 | - | 9 | 944 | |||
BC036828.1 | - | 9 | 945 | 5247772 | ||
M65066.1 | - | 8 | 837 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
