siRNA Details
Gene Symbol: BCL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000633.2 | NP_000624.2 | 3 | 1613 | ||
GenBank mRNA | BC027258.1 | - | 3 | 1509 | 4511027 | |
M13994.1 | - | 2 | 2576 | |||
M14745.1 | - | 2 | 1151 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
