siRNA Details
Gene Symbol: CENPBD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145039.3 | NP_659476.2 | 1 | 2599 | ||
GenBank mRNA | AK074351.1 | - | 2 | 1457 | ||
BC005105.1 | - | 2 | 632 | |||
BC040698.1 | - | 1 | 258 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
