siRNA Details
Gene Symbol: NMB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021077.3 | NP_066563.2 | 2 | 685 | ||
NM_205858.1 | NP_995580.1 | 2 | 685 | |||
GenBank mRNA | AL110112.1 | - | 2 | 304 | ||
BC007407.2 | - | 2 | 678 | 3030430 | ||
BC007431.2 | - | 2 | 678 | 3030430 | ||
BC008603.1 | - | 2 | 305 | 4185578 | ||
BI858080.1 | - | 2 | 638 | |||
M21551.1 | - | 2 | 302 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
