siRNA Details
Gene Symbol: SNTB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006750.3 | NP_006741.1 | 7 | 1635 | ||
GenBank mRNA | AF243385.1 | - | 6 | 1424 | ||
BC048215.1 | - | 7 | 1620 | 5805707 | ||
U40572.1 | - | 7 | 1635 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
