siRNA Details
Gene Symbol: WDHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008396.2 | NP_001008397.1 | 22 | 2772 | ||
NM_007086.3 | NP_009017.1 | 23 | 2865 | |||
XM_006720012.2 | XP_006720075.1 | 23 | 2857 | |||
XM_011536373.2 | XP_011534675.1 | 22 | 2664 | |||
XM_017020940.1 | XP_016876429.1 | 22 | 2770 | |||
XM_017020941.1 | XP_016876430.1 | 20 | 2468 | |||
GenBank mRNA | AJ006266.1 | - | 22 | 2814 | ||
AK001538.1 | - | 11 | 1419 | |||
AK001585.1 | - | 13 | 1788 | |||
AK001620.1 | - | 6 | 592 | |||
AK293005.1 | - | 23 | 2832 | |||
BC063041.1 | - | 23 | 2841 | 6514641 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
