siRNA Details
Gene Symbol: WDHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008396.2 | NP_001008397.1 | 11, 12 | 1341 | ||
NM_007086.3 | NP_009017.1 | 12, 13 | 1434 | |||
XM_006720012.2 | XP_006720075.1 | 12, 13 | 1432 | |||
XM_011536373.2 | XP_011534675.1 | 11, 12 | 1233 | |||
XM_017020940.1 | XP_016876429.1 | 11, 12 | 1345 | |||
XM_017020941.1 | XP_016876430.1 | 9, 10 | 1043 | |||
GenBank mRNA | AJ006266.1 | - | 11, 12 | 1383 | ||
AK001585.1 | - | 2, 3 | 357 | |||
AK293005.1 | - | 12, 13 | 1401 | |||
BC000622.2 | - | 12, 13 | 1399 | |||
BC005040.1 | - | 12, 13 | 1399 | |||
BC043349.1 | - | 12, 13 | 1422 | |||
BC063041.1 | - | 12, 13 | 1410 | 6514641 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
