siRNA Details
Gene Symbol: SIGLEC14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098612.1 | NP_001092082.1 | 5 | 898 | ||
GenBank mRNA | AK298284.1 | - | 5 | 820 | ||
AY854038.1 | - | 5 | 819 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
